Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGAGTCAGCCCACAGACTGGCTG[G/T]TCGCTTCACCTCCCTGGGGCTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126064 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHODH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHODH - dihydroorotate dehydrogenase (quinone) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001361.4 | 176 | Missense Mutation | GGT,GTT | G,V 59 | NP_001352.2 | |
XM_005255827.3 | 176 | Missense Mutation | GGT,GTT | G,V 31 | XP_005255884.1 | |
XM_005255829.3 | 176 | Intron | XP_005255886.1 | |||
XM_017022990.1 | 176 | UTR 5 | XP_016878479.1 |