Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAGTCCAGTTTGACGTCTGGAA[C/T]CTCATCTTCTGGCTCTGGATCCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611972 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPS5 - mitochondrial ribosomal protein S5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321995.1 | 1258 | Missense Mutation | ATT,GTT | I,V 322 | NP_001308924.1 | |
NM_001321996.1 | 1258 | Missense Mutation | ATT,GTT | I,V 235 | NP_001308925.1 | |
NM_001321997.1 | 1258 | Missense Mutation | ATT,GTT | I,V 235 | NP_001308926.1 | |
NM_031902.4 | 1258 | Missense Mutation | ATT,GTT | I,V 401 | NP_114108.1 |