Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTCAAACACACAGCCCCTGCT[A/T]CCCCTGGTGCTGCAGCGCCCTCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611239 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDHR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDHR2 - cadherin related family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPRIN1 - G protein regulated inducer of neurite outgrowth 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052899.2 | 2133 | Missense Mutation | GAA,GTA | E,V 653 | NP_443131.2 |