Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTGTCCCAAACACCAAGAAGC[C/T]CTGAAGCTCTTCTGCGAGGTAGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4638 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4638 - microRNA 4638 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM41 - tripartite motif containing 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033549.4 | 1434 | Silent Mutation | GCC,GCT | A,A 233 | NP_291027.3 | |
NM_201627.2 | 1434 | Silent Mutation | GCC,GCT | A,A 233 | NP_963921.1 | |
XM_006714929.2 | 1434 | Silent Mutation | GCC,GCT | A,A 233 | XP_006714992.1 | |
XM_006714930.2 | 1434 | Silent Mutation | GCC,GCT | A,A 233 | XP_006714993.1 | |
XM_006714931.3 | 1434 | Silent Mutation | GCC,GCT | A,A 233 | XP_006714994.1 | |
XM_006714932.3 | 1434 | Silent Mutation | GCC,GCT | A,A 233 | XP_006714995.1 |