Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTCTTGGTAGGAAGAAGATGCA[A/G]GCCAGCCCCATCCGTATCCCAACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606586 MIM: 616344 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAI14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAI14 - retinoic acid induced 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC23L - tetratricopeptide repeat domain 23 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317949.1 | 290 | Silent Mutation | CAA,CAG | Q,Q 11 | NP_001304878.1 | |
NM_144725.3 | 290 | Silent Mutation | CAA,CAG | Q,Q 2 | NP_653326.3 | |
XM_011513984.2 | 290 | Silent Mutation | CAA,CAG | Q,Q 2 | XP_011512286.1 | |
XM_011513985.2 | 290 | Silent Mutation | CAA,CAG | Q,Q 2 | XP_011512287.1 | |
XM_011513987.2 | 290 | Silent Mutation | CAA,CAG | Q,Q 2 | XP_011512289.1 | |
XM_017009119.1 | 290 | Silent Mutation | CAA,CAG | Q,Q 11 | XP_016864608.1 | |
XM_017009120.1 | 290 | Silent Mutation | CAA,CAG | Q,Q 2 | XP_016864609.1 | |
XM_017009121.1 | 290 | Silent Mutation | CAA,CAG | Q,Q 2 | XP_016864610.1 | |
XM_017009122.1 | 290 | Silent Mutation | CAA,CAG | Q,Q 2 | XP_016864611.1 | |
XM_017009123.1 | 290 | Silent Mutation | CAA,CAG | Q,Q 2 | XP_016864612.1 | |
XM_017009124.1 | 290 | Missense Mutation | AAG,AGG | K,R 10 | XP_016864613.1 | |
XM_017009125.1 | 290 | UTR 5 | XP_016864614.1 | |||
XM_017009126.1 | 290 | UTR 5 | XP_016864615.1 |