Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTCTCCGGCTCCTGCGAGCGCC[A/G]TCGGCGGGGCCAGCTCGCAGCCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615466 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MESDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MESDC1 - mesoderm development candidate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022566.2 | 1382 | Missense Mutation | ATC,GTC | I,V 22 | NP_072088.1 |
MIR4514 - microRNA 4514 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |