Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATCCTGGGTTAGACGGATACAA[A/G]AGCCTGGGAAGAATGGCCAGTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604812 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LILRA2 - leukocyte immunoglobulin like receptor A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130917.2 | 807 | Missense Mutation | AAG,GAG | K,E 75 | NP_001124389.2 | |
NM_001290270.1 | 807 | Missense Mutation | AAG,GAG | K,E 63 | NP_001277199.1 | |
NM_001290271.1 | 807 | Missense Mutation | AAG,GAG | K,E 75 | NP_001277200.1 | |
NM_006866.3 | 807 | Missense Mutation | AAG,GAG | K,E 75 | NP_006857.2 | |
XM_006722986.1 | 807 | Missense Mutation | AAG,GAG | K,E 75 | XP_006723049.1 | |
XM_011526385.1 | 807 | Missense Mutation | AAG,GAG | K,E 75 | XP_011524687.1 | |
XM_011526386.1 | 807 | Missense Mutation | AAG,GAG | K,E 63 | XP_011524688.1 | |
XM_011526387.1 | 807 | Missense Mutation | AAG,GAG | K,E 63 | XP_011524689.1 | |
XM_011526388.1 | 807 | Missense Mutation | AAG,GAG | K,E 63 | XP_011524690.1 | |
XM_011526389.1 | 807 | Intron | XP_011524691.1 | |||
XM_011526390.1 | 807 | Missense Mutation | AAG,GAG | K,E 75 | XP_011524692.1 | |
XM_011526391.1 | 807 | Missense Mutation | AAG,GAG | K,E 75 | XP_011524693.1 | |
XM_017026224.1 | 807 | Missense Mutation | AAG,GAG | K,E 75 | XP_016881713.1 |