Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCCTCTGCAGCCCAAGCCGGTC[C/T]GCGTTCTTGACGGGAAGATACCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300003 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARSF - arylsulfatase F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201538.1 | 385 | Silent Mutation | TCC,TCT | S,S 83 | NP_001188467.1 | |
NM_001201539.1 | 385 | Silent Mutation | TCC,TCT | S,S 83 | NP_001188468.1 | |
NM_004042.4 | 385 | Intron | NP_004033.2 | |||
XM_011545522.1 | 385 | Silent Mutation | TCC,TCT | S,S 100 | XP_011543824.1 | |
XM_011545523.1 | 385 | Silent Mutation | TCC,TCT | S,S 100 | XP_011543825.1 | |
XM_011545524.1 | 385 | Silent Mutation | TCC,TCT | S,S 100 | XP_011543826.1 | |
XM_017029527.1 | 385 | Silent Mutation | TCC,TCT | S,S 83 | XP_016885016.1 | |
XM_017029528.1 | 385 | Silent Mutation | TCC,TCT | S,S 100 | XP_016885017.1 |