Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGATGCGCTGGTGGCGGATGAG[C/G]GCTGAGGAGAAGCGGAAGGCCTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMIM17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMIM17 - small integral membrane protein 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF835 - zinc finger protein 835 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005850.2 | 1054 | Silent Mutation | GCC,GCG | A,A 264 | NP_001005850.2 | |
XM_005259382.2 | 1054 | Silent Mutation | GCC,GCG | A,A 264 | XP_005259439.1 | |
XM_005259383.3 | 1054 | Silent Mutation | GCC,GCG | A,A 264 | XP_005259440.1 |