Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTGAGTTCACAGCCACCATCTC[C/G]TCCTGCCCCAATATGCTGTACCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAT1 - adenosine deaminase, tRNA specific 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324444.1 | Intron | NP_001311373.1 | ||||
NM_001324445.1 | Intron | NP_001311374.1 | ||||
NM_001324446.1 | Intron | NP_001311375.1 | ||||
NM_001324448.1 | Intron | NP_001311377.1 | ||||
NM_001324449.1 | Intron | NP_001311378.1 | ||||
NM_001324450.1 | Intron | NP_001311379.1 | ||||
NM_001324451.1 | Intron | NP_001311380.1 | ||||
NM_001324452.1 | Intron | NP_001311381.1 | ||||
NM_001324453.1 | Intron | NP_001311382.1 | ||||
NM_012091.4 | Intron | NP_036223.2 |