Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGACCAGGAGGACCTTGTTCTCC[A/C]GGATACCCAAACCCTTGGCTCCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610002 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL21A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL21A1 - collagen type XXI alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318751.1 | 3020 | Silent Mutation | CCG,CCT | P,P 886 | NP_001305680.1 | |
NM_001318752.1 | 3020 | Silent Mutation | CCG,CCT | P,P 883 | NP_001305681.1 | |
NM_001318753.1 | 3020 | Silent Mutation | CCG,CCT | P,P 284 | NP_001305682.1 | |
NM_001318754.1 | 3020 | Silent Mutation | CCG,CCT | P,P 243 | NP_001305683.1 | |
NM_030820.3 | 3020 | Silent Mutation | CCG,CCT | P,P 886 | NP_110447.2 | |
XM_006715223.1 | 3020 | Silent Mutation | CCG,CCT | P,P 874 | XP_006715286.1 | |
XM_011514924.2 | 3020 | Silent Mutation | CCG,CCT | P,P 886 | XP_011513226.1 | |
XM_011514925.2 | 3020 | Silent Mutation | CCG,CCT | P,P 886 | XP_011513227.1 | |
XM_011514926.1 | 3020 | Silent Mutation | CCG,CCT | P,P 886 | XP_011513228.1 | |
XM_011514927.1 | 3020 | Silent Mutation | CCG,CCT | P,P 886 | XP_011513229.1 |