Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCGCAGCAGCGTCCCGCGAGG[C/T]CTCGTGTGGCCGAAGTCGGTCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SBK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SBK2 - SH3 domain binding kinase family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101401.2 | 639 | Silent Mutation | AGA,AGG | R,R 208 | NP_001094871.2 | |
XM_006723327.3 | 639 | Silent Mutation | AGA,AGG | R,R 208 | XP_006723390.1 | |
XM_011527227.2 | 639 | Intron | XP_011525529.1 |