Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGAACCAAGTGGATATGCTGCT[C/G]CAGGAAATGGCAGATGAGGCGGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606486 MIM: 139312 | ||||||||||||||||||||
Literature Links: |
CHMP1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHMP1B - charged multivesicular body protein 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020412.4 | 581 | Silent Mutation | CTC,CTG | L,L 153 | NP_065145.2 |
GNAL - G protein subunit alpha L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142339.2 | 581 | Intron | NP_001135811.1 | |||
NM_001261443.1 | 581 | Intron | NP_001248372.1 | |||
NM_001261444.1 | 581 | Intron | NP_001248373.1 | |||
NM_182978.3 | 581 | Intron | NP_892023.1 | |||
XM_006722324.2 | 581 | Intron | XP_006722387.1 |