Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGATGCGCTCTCGTACATTTCTC[A/G]GCTGGTACTGTAATTTGAAGGGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612208 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLPH3L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GOLPH3L - golgi phosphoprotein 3 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018178.5 | 611 | Missense Mutation | CCG,CTG | P,L 156 | NP_060648.2 | |
XM_006711428.1 | 611 | Missense Mutation | CCG,CTG | P,L 170 | XP_006711491.1 |