Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGACTGCAGGAAGAGCAGCGAGCC[A/C]GGCTCGGCGCCCAGCGACAGGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609895 MIM: 601791 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASZ1 - castor zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079843.2 | 5498 | Silent Mutation | NP_001073312.1 | |||
NM_017766.4 | 5498 | Intron | NP_060236.3 | |||
XM_005263479.3 | 5498 | Silent Mutation | XP_005263536.1 | |||
XM_011541635.2 | 5498 | Intron | XP_011539937.1 | |||
XM_017001539.1 | 5498 | Silent Mutation | XP_016857028.1 | |||
XM_017001540.1 | 5498 | Silent Mutation | XP_016857029.1 | |||
XM_017001541.1 | 5498 | Silent Mutation | XP_016857030.1 |
PEX14 - peroxisomal biogenesis factor 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |