Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCACCACCCTCTGAGCACCCTCA[C/T]TGGGTTGGGTGGGGGCCAAAGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610934 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOBOX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOBOX - NOBOX oogenesis homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080413.3 | 1407 | Missense Mutation | AAT,AGT | N,S 431 | NP_001073882.3 | |
XM_017011742.1 | 1407 | Missense Mutation | AAT,AGT | N,S 399 | XP_016867231.1 |