Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTGGAGGCTGGGAACCCCGCAG[C/T]GCGAGGCTGCCTCATCAACGGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609023 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATIP-AS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CATIP-AS2 - CATIP antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6810 - microRNA 6810 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNKD - paroxysmal nonkinesigenic dyskinesia | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077399.2 | 1513 | Intron | NP_001070867.1 | |||
NM_015488.4 | 1513 | UTR 3 | NP_056303.3 | |||
NM_022572.4 | 1513 | UTR 3 | NP_072094.1 | |||
XM_017003771.1 | 1513 | UTR 3 | XP_016859260.1 | |||
XM_017003772.1 | 1513 | UTR 3 | XP_016859261.1 |