Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATGATTACAGGAAAGCTGGTAG[A/G]TGTTTTCAATGAGCCCTTCTTCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603028 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF175 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF175 - ring finger protein 175 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173662.2 | 1197 | Missense Mutation | ACC,ATC | T,I 247 | NP_775933.1 | |
XM_005262938.3 | 1197 | Missense Mutation | ACC,ATC | T,I 247 | XP_005262995.1 | |
XM_005262939.3 | 1197 | Missense Mutation | ACC,ATC | T,I 167 | XP_005262996.1 | |
XM_005262940.4 | 1197 | Missense Mutation | ACC,ATC | T,I 166 | XP_005262997.1 | |
XM_011531879.2 | 1197 | Missense Mutation | ACC,ATC | T,I 215 | XP_011530181.1 | |
XM_011531881.2 | 1197 | Missense Mutation | ACC,ATC | T,I 207 | XP_011530183.1 | |
XM_011531882.2 | 1197 | Missense Mutation | ACC,ATC | T,I 247 | XP_011530184.1 | |
XM_011531883.2 | 1197 | Missense Mutation | ACC,ATC | T,I 175 | XP_011530185.1 | |
XM_017008047.1 | 1197 | Missense Mutation | ACC,ATC | T,I 148 | XP_016863536.1 |
TLR2 - toll like receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |