Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACTTGATGATGCGGGCGTTCC[G/T]CTCGATGACCGAAGTGGTGCTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611395 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM110C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM110C - family with sequence similarity 110 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077710.2 | 1010 | Silent Mutation | AGG,CGG | R,R 289 | NP_001071178.2 | |
XM_011510372.2 | 1010 | Silent Mutation | AGG,CGG | R,R 289 | XP_011508674.1 | |
XM_017004689.1 | 1010 | Silent Mutation | AGG,CGG | R,R 289 | XP_016860178.1 | |
XM_017004690.1 | 1010 | Silent Mutation | AGG,CGG | R,R 289 | XP_016860179.1 | |
XM_017004691.1 | 1010 | Silent Mutation | AGG,CGG | R,R 289 | XP_016860180.1 | |
XM_017004692.1 | 1010 | Silent Mutation | AGG,CGG | R,R 289 | XP_016860181.1 |