Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGCTGGGCCCGCAGGGCCTCCA[C/T]GTCCCCCTCCAGCTCCACGGCCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617003 MIM: 615403 MIM: 605914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BICDL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BICDL2 - BICD family like cargo adaptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001103175.1 | 479 | Missense Mutation | ATG,GTG | M,V 125 | NP_001096645.1 | |
XM_005255135.4 | 479 | Missense Mutation | ATG,GTG | M,V 125 | XP_005255192.1 | |
XM_011522391.1 | 479 | Missense Mutation | ATG,GTG | M,V 60 | XP_011520693.1 |
HCFC1R1 - host cell factor C1 regulator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100128770 - uncharacterized LOC100128770 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THOC6 - THO complex 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFRSF12A - TNF receptor superfamily member 12A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |