Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAAAAGAGAAATACCTGCATTG[C/T]GCCACAATTACTTCCTTGAGGCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608418 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNI2 - cyclin I family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039780.3 | 679 | Silent Mutation | TGC,TGT | C,C 194 | NP_001034869.1 | |
NM_001287252.1 | 679 | Silent Mutation | TGC,TGT | C,C 194 | NP_001274181.1 | |
NM_001287253.1 | 679 | Silent Mutation | TGC,TGT | C,C 195 | NP_001274182.1 | |
XM_017009725.1 | 679 | Silent Mutation | TGC,TGT | C,C 195 | XP_016865214.1 |
SEPT8 - septin 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |