Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCCCCCGCGCAGTCACCACGAC[A/G]CGGCACCCGTGGACGGGCGTTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609607 MIM: 613466 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHDC9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHDC9 - kelch domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007255.2 | 395 | Missense Mutation | ACG,GCG | T,A 84 | NP_001007256.1 | |
NM_152366.4 | 395 | Missense Mutation | ACG,GCG | T,A 84 | NP_689579.3 |
NECTIN4 - nectin cell adhesion molecule 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFDN2 - prefoldin subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |