Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGATGAAGGGGAATTCACCTGT[C/T]GAGCTCAGAACTCTCTGGGTTCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604410 MIM: 605640 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928517 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928517 - uncharacterized LOC101928517 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGLEC7 - sialic acid binding Ig like lectin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277201.1 | 1030 | Intron | NP_001264130.1 | |||
NM_014385.3 | 1030 | Nonsense Mutation | CGA,TGA | R,* 321 | NP_055200.1 | |
NM_016543.3 | 1030 | Nonsense Mutation | CGA,TGA | R,* 228 | NP_057627.2 | |
XM_011526721.2 | 1030 | Nonsense Mutation | CGA,TGA | R,* 212 | XP_011525023.1 |
SIGLEC9 - sialic acid binding Ig like lectin 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |