Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGTAAACAACAAAACGAAGTTT[C/T]GTCTTGTATCCCCAAAATATTAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613429 MIM: 614642 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAUS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAUS2 - HAUS augmin like complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130447.1 | 560 | Missense Mutation | TCG,TTG | S,L 167 | NP_001123919.1 | |
NM_001323629.1 | 560 | Intron | NP_001310558.1 | |||
NM_001323630.1 | 560 | Missense Mutation | TCG,TTG | S,L 67 | NP_001310559.1 | |
NM_001323631.1 | 560 | Missense Mutation | TCG,TTG | S,L 104 | NP_001310560.1 | |
NM_001323632.1 | 560 | Missense Mutation | TCG,TTG | S,L 104 | NP_001310561.1 | |
NM_018097.2 | 560 | Missense Mutation | TCG,TTG | S,L 198 | NP_060567.1 | |
XM_017022396.1 | 560 | Intron | XP_016877885.1 | |||
XM_017022397.1 | 560 | Intron | XP_016877886.1 |
STARD9 - StAR related lipid transfer domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |