Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTAAAAATCCTTTATAAAAGTGG[G/T]ATTATATGGCTATCCTGGTATTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609335 MIM: 604301 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf27 - chromosome 1 open reading frame 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164245.1 | 564 | Intron | NP_001157717.1 | |||
NM_001164246.1 | 564 | Intron | NP_001157718.1 | |||
NM_017847.5 | 564 | Intron | NP_060317.3 | |||
XM_011509660.1 | 564 | Intron | XP_011507962.1 | |||
XM_011509661.1 | 564 | Intron | XP_011507963.1 | |||
XM_017001543.1 | 564 | Intron | XP_016857032.1 |
OCLM - oculomedin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022375.3 | 564 | Silent Mutation | GGG,GGT | G,G 30 | NP_071770.1 |