Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATATTGTGGAATGGAACGTACA[C/G]ACATGCAAAGTAAAGTGGTGAGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616195 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNORD92 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNORD92 - small nucleolar RNA, C/D box 92 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR43 - WD repeat domain 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015131.2 | 524 | Missense Mutation | CAC,CAG | H,Q 156 | NP_055946.1 |