Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTACATCCTTAGCCTCATCCGAA[A/G]AAAGGCCAGATGAACCAGGTCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102910 MIM: 605682 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5B - ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001686.3 | Intron | NP_001677.2 |
BAZ2A - bromodomain adjacent to zinc finger domain 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD59A - small nucleolar RNA, C/D box 59A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD59B - small nucleolar RNA, C/D box 59B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |