Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGGAGAGCGACATCACGCGAC[A/G]TAGCTGTATTCATCTGCCGACGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603135 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUL2 - cullin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198777.1 | 2624 | Silent Mutation | TAC,TAT | Y,Y 743 | NP_001185706.1 | |
NM_001198778.1 | 2624 | Silent Mutation | TAC,TAT | Y,Y 762 | NP_001185707.1 | |
NM_001198779.1 | 2624 | Silent Mutation | TAC,TAT | Y,Y 756 | NP_001185708.1 | |
NM_001324375.1 | 2624 | Silent Mutation | TAC,TAT | Y,Y 680 | NP_001311304.1 | |
NM_001324376.1 | 2624 | Silent Mutation | TAC,TAT | Y,Y 612 | NP_001311305.1 | |
NM_003591.3 | 2624 | Silent Mutation | TAC,TAT | Y,Y 743 | NP_003582.2 | |
XM_011519743.1 | 2624 | Silent Mutation | TAC,TAT | Y,Y 806 | XP_011518045.1 | |
XM_011519744.1 | 2624 | Silent Mutation | TAC,TAT | Y,Y 806 | XP_011518046.1 | |
XM_011519745.1 | 2624 | Silent Mutation | TAC,TAT | Y,Y 806 | XP_011518047.1 | |
XM_011519747.1 | 2624 | Silent Mutation | TAC,TAT | Y,Y 743 | XP_011518049.1 |