Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCGGAGTTCCTGGAAGACCCCTC[G/T]GTCCTGACAAAAGACAAGTTGAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 188380 | ||||||||||||||||||||
Literature Links: |
TMPO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMPO - thymopoietin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032283.2 | 313 | Silent Mutation | TCG,TCT | S,S 9 | NP_001027454.1 | |
NM_001032284.2 | 313 | Silent Mutation | TCG,TCT | S,S 9 | NP_001027455.1 | |
NM_001307975.1 | 313 | Silent Mutation | TCG,TCT | S,S 9 | NP_001294904.1 | |
NM_003276.2 | 313 | Silent Mutation | TCG,TCT | S,S 9 | NP_003267.1 | |
XM_005269132.3 | 313 | Silent Mutation | TCG,TCT | S,S 9 | XP_005269189.1 | |
XM_017019914.1 | 313 | Intron | XP_016875403.1 |
TMPO-AS1 - TMPO antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |