Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTGTGTCCACACAGGTCAGGGC[T/G]ACCCTTCAGTTTACCTGAGAAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605089 MIM: 601754 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf39 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C22orf39 - chromosome 22 open reading frame 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166242.1 | 1204 | Intron | NP_001159714.1 | |||
NM_173793.4 | 1204 | UTR 3 | NP_776154.3 |
MRPL40 - mitochondrial ribosomal protein L40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFD1L - ubiquitin fusion degradation 1 like (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |