Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGCCTGCTTGGACCTGCACATG[C/T]GCTTGTACTGTCCCCTGAAGGATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAMD9L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAMD9L - sterile alpha motif domain containing 9 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303496.1 | 4432 | Missense Mutation | CAC,CGC | H,R 1465 | NP_001290425.1 | |
NM_001303497.1 | 4432 | Missense Mutation | CAC,CGC | H,R 1465 | NP_001290426.1 | |
NM_001303498.1 | 4432 | Missense Mutation | CAC,CGC | H,R 1465 | NP_001290427.1 | |
NM_001303500.1 | 4432 | Missense Mutation | CAC,CGC | H,R 1465 | NP_001290429.1 | |
NM_152703.3 | 4432 | Missense Mutation | CAC,CGC | H,R 1465 | NP_689916.2 | |
XM_005250193.3 | 4432 | Missense Mutation | CAC,CGC | H,R 1465 | XP_005250250.1 | |
XM_006715890.1 | 4432 | Missense Mutation | CAC,CGC | H,R 1465 | XP_006715953.1 | |
XM_011515903.2 | 4432 | Missense Mutation | CAC,CGC | H,R 1465 | XP_011514205.1 | |
XM_017011822.1 | 4432 | Missense Mutation | CAC,CGC | H,R 1465 | XP_016867311.1 | |
XM_017011823.1 | 4432 | Missense Mutation | CAC,CGC | H,R 1465 | XP_016867312.1 |