Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGCTGGTATTTCTCCGGCTTGC[A/G]GACGTGCAGGATCTCCACGTTGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601041 MIM: 612399 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TLE2 - transducin like enhancer of split 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144761.1 | 2278 | Missense Mutation | CGC,TGC | R,C 674 | NP_001138233.1 | |
NM_001144762.1 | 2278 | Missense Mutation | CGC,TGC | R,C 538 | NP_001138234.1 | |
NM_001300846.1 | 2278 | Missense Mutation | CGC,TGC | R,C 661 | NP_001287775.1 | |
NM_003260.4 | 2278 | Missense Mutation | CGC,TGC | R,C 660 | NP_003251.2 | |
XM_006722864.2 | 2278 | Missense Mutation | CGC,TGC | R,C 661 | XP_006722927.1 | |
XM_011528230.1 | 2278 | Missense Mutation | CGC,TGC | R,C 649 | XP_011526532.1 |
TLE6 - transducin like enhancer of split 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |