Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGGGTAGGTGAGTCCCCGAAGG[C/T]CCCCAGCACAGTGACTCCAGCCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616065 MIM: 609951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIANP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIANP - PILR alpha associated neural protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244014.1 | 1060 | Missense Mutation | ACC,GCC | T,A 241 | NP_001230943.1 | |
NM_001244015.1 | 1060 | Missense Mutation | ACC,GCC | T,A 241 | NP_001230944.1 | |
NM_153685.3 | 1060 | Missense Mutation | ACC,GCC | T,A 241 | NP_710152.1 | |
XM_011520926.2 | 1060 | Missense Mutation | ACC,GCC | T,A 241 | XP_011519228.1 |
ZNF384 - zinc finger protein 384 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |