Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCAGTTTCTTCTATAAGGTTCAC[A/G]ATCTAGAGGAGTGAAGAAGAGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159558 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLLT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MLLT3 - MLLT3, super elongation complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286691.1 | 1632 | Silent Mutation | ATC,ATT | I,I 523 | NP_001273620.1 | |
NM_004529.3 | 1632 | Silent Mutation | ATC,ATT | I,I 526 | NP_004520.2 | |
XM_017014726.1 | 1632 | Silent Mutation | ATC,ATT | I,I 478 | XP_016870215.1 | |
XM_017014727.1 | 1632 | Silent Mutation | ATC,ATT | I,I 475 | XP_016870216.1 |