Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATTTTGATATCTATCTTCATCA[C/T]TTCTTGAGCCTGAGTGCTTTCTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615186 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CWC22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CWC22 - CWC22 homolog, spliceosome-associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020943.2 | 2897 | Missense Mutation | AAT,AGT | N,S 866 | NP_065994.1 | |
XM_005246726.2 | 2897 | Missense Mutation | AAT,AGT | N,S 866 | XP_005246783.1 |