Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCCGCGGAGGCCGCTGCTTCA[G/T]CCCCTGCCCATCCTGCTCCTTGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600466 MIM: 608204 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH3B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH3B1 - aldehyde dehydrogenase 3 family member B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC93B1 - unc-93 homolog B1 (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030930.3 | 302 | Missense Mutation | ATG,CTG | M,L 204 | NP_112192.2 | |
XM_011545290.1 | 302 | Missense Mutation | ATG,CTG | M,L 67 | XP_011543592.1 | |
XM_011545291.2 | 302 | Missense Mutation | ATG,CTG | M,L 19 | XP_011543593.1 |