Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCGAAACTCATTTGCCAAGTTC[C/T]GGGTGTGTCTTGGAACAAACGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607587 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEF1AKMT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EEF1AKMT1 - eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318939.1 | 702 | Missense Mutation | CAG,CGG | Q,R 194 | NP_001305868.1 | |
NM_174928.2 | 702 | Missense Mutation | CAG,CGG | Q,R 194 | NP_777588.1 | |
XM_017020432.1 | 702 | Missense Mutation | CAG,CGG | Q,R 194 | XP_016875921.1 |
IL17D - interleukin 17D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |