Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCATAGTTCATTAATCCAGGTAC[A/G]CAAATACTTCTATCCCAATCCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608107 MIM: 603231 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEFV PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEFV - Mediterranean fever | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000243.2 | 2813 | UTR 3 | NP_000234.1 | |||
NM_001198536.1 | 2813 | UTR 3 | NP_001185465.1 | |||
XM_017023236.1 | 2813 | Intron | XP_016878725.1 |
ZNF200 - zinc finger protein 200 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |