Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCCATAATGAAGATGGAAGAAA[C/T]GGAGATGTCTCTCAGTTTCCATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613413 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM106B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM106B - transmembrane protein 106B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134232.1 | 332 | Silent Mutation | AAC,AAT | N,N 42 | NP_001127704.1 | |
NM_018374.3 | 332 | Silent Mutation | AAC,AAT | N,N 42 | NP_060844.2 | |
XM_005249789.1 | 332 | Silent Mutation | AAC,AAT | N,N 42 | XP_005249846.1 |