Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGGGAAGCAGCAGTGGAAGGGG[A/T]AGAGGAGGCTGTGTCCTATGGAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613799 MIM: 616637 | ||||||||||||||||||||
Literature Links: |
CCDC40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC40 - coiled-coil domain containing 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243342.1 | 270 | Missense Mutation | GAA,GTA | E,V 80 | NP_001230271.1 | |
NM_017950.3 | 270 | Missense Mutation | GAA,GTA | E,V 80 | NP_060420.2 | |
XM_005257492.4 | 270 | Missense Mutation | GAA,GTA | E,V 80 | XP_005257549.1 | |
XM_011524963.2 | 270 | Missense Mutation | GAA,GTA | E,V 50 | XP_011523265.1 | |
XM_011524964.2 | 270 | Intron | XP_011523266.1 | |||
XM_011524965.2 | 270 | Missense Mutation | GAA,GTA | E,V 80 | XP_011523267.1 | |
XM_017024807.1 | 270 | Missense Mutation | GAA,GTA | E,V 80 | XP_016880296.1 |
TBC1D16 - TBC1 domain family member 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |