Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGAGGGACATCAAAATTACTTTC[A/G]TCACTGCTCCTATCTGATTCTGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOP2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOP2A - topoisomerase (DNA) II alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001067.3 | 4013 | Silent Mutation | GAC,GAT | D,D 1304 | NP_001058.2 | |
XM_005257632.1 | 4013 | Silent Mutation | GAC,GAT | D,D 1292 | XP_005257689.1 | |
XM_011525165.1 | 4013 | Silent Mutation | GAC,GAT | D,D 1304 | XP_011523467.1 |