Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACCCGGAGGTTCTGCAGGGCCA[C/T]GTCGGCGTTCTGCTTCACCACGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613036 MIM: 602777 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PMPCA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PMPCA - peptidase, mitochondrial processing alpha subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SDCCAG3 - serologically defined colon cancer antigen 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039707.1 | 1145 | Intron | NP_001034796.1 | |||
NM_001039708.1 | 1145 | Intron | NP_001034797.1 | |||
NM_006643.3 | 1145 | Intron | NP_006634.3 | |||
XM_005266050.3 | 1145 | Intron | XP_005266107.1 | |||
XM_005266051.3 | 1145 | Intron | XP_005266108.1 | |||
XM_011518157.2 | 1145 | Missense Mutation | ATG,GTG | M,V 353 | XP_011516459.1 | |
XM_017014218.1 | 1145 | Intron | XP_016869707.1 | |||
XM_017014219.1 | 1145 | Intron | XP_016869708.1 |
SNAPC4 - small nuclear RNA activating complex polypeptide 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |