Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGAAAGACCGATACATGTAAAGG[A/G]TGTGGTTTGGAAACCAACAGAAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162341 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMBR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NMBR - neuromedin B receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324307.1 | 528 | Missense Mutation | AAC,AGC | N,S 136 | NP_001311236.1 | |
NM_001324308.1 | 528 | Missense Mutation | AAC,AGC | N,S 136 | NP_001311237.1 | |
NM_002511.3 | 528 | Missense Mutation | AAC,AGC | N,S 284 | NP_002502.2 | |
XM_011535846.1 | 528 | Missense Mutation | AAC,AGC | N,S 177 | XP_011534148.1 | |
XM_017010901.1 | 528 | Missense Mutation | AAC,AGC | N,S 195 | XP_016866390.1 | |
XM_017010902.1 | 528 | Missense Mutation | AAC,AGC | N,S 136 | XP_016866391.1 |