Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGTGTTCTAGGAATCTATAACA[C/T]TGAAGGATGTAGCTGTGAACTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF483 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF483 - zinc finger protein 483 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007169.3 | 746 | Silent Mutation | CTG,TTG | L,L 172 | NP_001007170.1 | |
NM_133464.3 | 746 | Silent Mutation | CTG,TTG | L,L 172 | NP_597721.2 | |
XM_011518300.2 | 746 | Silent Mutation | CTG,TTG | L,L 172 | XP_011516602.1 | |
XM_017014337.1 | 746 | Silent Mutation | CTG,TTG | L,L 172 | XP_016869826.1 | |
XM_017014338.1 | 746 | Silent Mutation | CTG,TTG | L,L 172 | XP_016869827.1 | |
XM_017014339.1 | 746 | Silent Mutation | CTG,TTG | L,L 172 | XP_016869828.1 |