Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCCTGGCCTCCAGTTTCTGGAT[C/G]TGCTTCTTCCCACCCTTCAGCGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160741 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYH8 - myosin heavy chain 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002472.2 | 5537 | Silent Mutation | CAC,CAG | H,Q 1814 | NP_002463.2 |
MYHAS - myosin heavy chain gene cluster antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |