Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCTACCAGTAAAATATACTTAC[A/C]AGAGTCTTCTAGAGAAGGCCAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614406 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130950 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100130950 - uncharacterized LOC100130950 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCIMP - SLP adaptor and CSK interacting membrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271842.1 | 388 | Missense Mutation | GCC,TCC | A,S 88 | NP_001258771.1 | |
NM_001319190.1 | 388 | Missense Mutation | GAT,TAT | D,Y 88 | NP_001306119.1 | |
NM_207103.3 | 388 | Missense Mutation | GCC,TCC | A,S 95 | NP_996986.1 | |
XM_017024615.1 | 388 | Missense Mutation | GAT,TAT | D,Y 95 | XP_016880104.1 |