Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACACCATCCTTGGCTTTACTAC[A/G]ATTTGTGCCTGCTTCTCAATTAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616934 | ||||||||||||||||||||
Literature Links: |
CCDC43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC43 - coiled-coil domain containing 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099225.1 | 383 | Silent Mutation | ATC,ATT | I,I 113 | NP_001092695.1 | |
NM_144609.2 | 383 | Silent Mutation | ATC,ATT | I,I 113 | NP_653210.2 |
MEIOC - meiosis specific with coiled-coil domain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |