Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAAGAAACTGGGAAGCTCCACT[C/T]CCCACCGCCTCTGCCTGGGTGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 136950 | ||||||||||||||||||||
Literature Links: |
FURIN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FURIN - furin, paired basic amino acid cleaving enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289823.1 | Intron | NP_001276752.1 | ||||
NM_001289824.1 | Intron | NP_001276753.1 | ||||
NM_002569.3 | Intron | NP_002560.1 |