Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCATTTGGGCCTTCAGTGGGTC[A/G]TCCACCTCCACATCGATGTCGTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601681 MIM: 601736 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FTSJ3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FTSJ3 - FtsJ homolog 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMC5 - proteasome 26S subunit, ATPase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMARCD2 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098426.1 | 1103 | Silent Mutation | GAC,GAT | D,D 413 | NP_001091896.1 | |
XM_005257604.2 | 1103 | Silent Mutation | GAC,GAT | D,D 338 | XP_005257661.2 | |
XM_017024967.1 | 1103 | Silent Mutation | GAC,GAT | D,D 365 | XP_016880456.1 |