Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGATGCCTTATGGCACTGGACT[A/G]ACCCCACAGCCTATTCAGAACACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602017 MIM: 600075 | ||||||||||||||||||||
Literature Links: |
PSMB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSMB1 - proteasome subunit beta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBP - TATA-box binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172085.1 | 191 | Silent Mutation | CTA,CTG | L,L 18 | NP_001165556.1 | |
NM_003194.4 | 191 | Silent Mutation | CTA,CTG | L,L 38 | NP_003185.1 |